Learn More
Bioworld Luria-Bertani (LB) Broth, Lennox, 5 kg
Catalog No. 50254319
Encompass_Preferred
Luria-Bertani (LB) Broth, LennoxFor propagation and maintenance of Escherichia coliLow salt formulationpH: 7.0Working Concentration: 20 g/LDehydrated Appearance: light beige, homogeneous powderPrepared Appearance: light amber brothSoluble in waterContains:TryptoneYeastSodium Choride Usually ships within 24 hours. This product is sold for laboratory research use only.
Catalog No. 50-254-319 Supplier Bioworld Supplier No. 306200383
Luria-Bertani (LB) Broth, LennoxFor propagation and maintenance of Escherichia coliLow salt formulationpH: 7.0Working Concentration: 20 g/LDehydrated Appearance: light beige, homogeneous powderPrepared Appearance: light amber brothSoluble in waterContains:TryptoneYeastSodium Choride Usually ships within 24 hours. This product is sold for laboratory research use only.