Learn More
Bioworld Luria-Bertani (LB) Broth, Miller, 5 kg
Catalog No. 50254326
Encompass_Preferred
Luria-Bertani (LB) Broth, MillerFor propagation and maintenance of Escherichia coliHigh salt formulationpH: 7.0Working Concentration: 25 g/LDehydrated Appearance: light tan, homogeneous powderPrepared Appearance: light amber brothSoluble in waterContains (g/L):Tryptone-10gYeast -5gSodium Choride -10gUsually ships within 24 hours.This product is sold for laboratory research use only.
Catalog No. 50-254-326 Supplier Bioworld Supplier No. 306200443
Luria-Bertani (LB) Broth, MillerFor propagation and maintenance of Escherichia coliHigh salt formulationpH: 7.0Working Concentration: 25 g/LDehydrated Appearance: light tan, homogeneous powderPrepared Appearance: light amber brothSoluble in waterContains (g/L):Tryptone-10gYeast -5gSodium Choride -10gUsually ships within 24 hours.This product is sold for laboratory research use only.