Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quality Biological Inc LB Agar Plates, 1.5% (Lennox), Sleeve 10 X 25 mL
SDP

Catalog No. 507517574
Encompass_Preferred

LB Agar 1.5% is useful for propagation of bacteriological stocks, as well as plating and screening bacteriological clones. LB Agar 1.5% Lennox contains half the sodium chloride level of the Miller formulation. LB Agar 1.5% is useful for propagation of bacteriological stocks, as well as plating and screening bacteriological clones.

Just autoclave to melt, and pour.

Storage Conditions: 15°C to 30°C
Shipping Condition: ambient temperature
Autoclave Sterilized
Max Shelf Life: 6 months from date of manufacture

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Catalog No. 50-751-7574 Supplier Quality Biological Inc Supplier No. 340025231
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

LB Agar 1.5% is useful for propagation of bacteriological stocks, as well as plating and screening bacteriological clones. LB Agar 1.5% Lennox contains half the sodium chloride level of the Miller formulation. LB Agar 1.5% is useful for propagation of bacteriological stocks, as well as plating and screening bacteriological clones.

Just autoclave to melt, and pour.

Storage Conditions: 15°C to 30°C
Shipping Condition: ambient temperature
Autoclave Sterilized
Max Shelf Life: 6 months from date of manufacture

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.