Learn More
Bioworld Luria Broth Base, Miller, 500 g
Catalog No. 50254432
Encompass_Preferred
Luria Broth BaseFormulated for the propagation and maintenance of E. coli for molecular biologyContains one-tenth the sodium chloride content as LB Broth, Lennox and one-twentieth the sodium chloride content as LB Broth, Miller formulationspH: 7.0Dehydrated Appearance: light tan, homogeneous powderPrepared Appearance: light amber brothSoluble in boiling waterComposition (g/L):Bacto Tryptone- 10g Bacto Yeast Extract-5gSodium Chloride-0.5g
Catalog No. 50-254-432 Supplier Bioworld Supplier No. 306210131
Luria Broth BaseFormulated for the propagation and maintenance of E. coli for molecular biologyContains one-tenth the sodium chloride content as LB Broth, Lennox and one-twentieth the sodium chloride content as LB Broth, Miller formulationspH: 7.0Dehydrated Appearance: light tan, homogeneous powderPrepared Appearance: light amber brothSoluble in boiling waterComposition (g/L):Bacto Tryptone- 10g Bacto Yeast Extract-5gSodium Chloride-0.5g