Learn More
AG Scientific Inc Luria Broth Capsules 500 G
Catalog No. 502268321
Encompass_Preferred
Luria Broth is now available in convenient pre-measured and pre-mixed capsules. Prebuffered media eliminates messy weighing and pH adjustments. Molecular biology grade.For maintenance and propagation of E. Coli.Protocol: Simply dissolve 25 capsules per 1 Liter of distilled water. Water must be 37°C or warmer to completely dissolve gelatin capsule. Agitate. Sterilize by autoclaving for 15 minutes before use. Buffer is set for 7.0 ± 0.2 at 25°C using a 10mM solution of Tris/Tris HCl. pH will vary depending on temperature.
Catalog No. 50-226-8321 Supplier AG Scientific Inc Supplier No. L2596500GM
Luria Broth is now available in convenient pre-measured and pre-mixed capsules. Prebuffered media eliminates messy weighing and pH adjustments. Molecular biology grade.For maintenance and propagation of E. Coli.Protocol: Simply dissolve 25 capsules per 1 Liter of distilled water. Water must be 37°C or warmer to completely dissolve gelatin capsule. Agitate. Sterilize by autoclaving for 15 minutes before use. Buffer is set for 7.0 ± 0.2 at 25°C using a 10mM solution of Tris/Tris HCl. pH will vary depending on temperature.