Learn More
Alkali Scientific Luria Broth, High Salt Formula, Powder [Miller's LB Broth], 10 Kg
SDP
Catalog No. 5000660578
Encompass_Preferred
Luria Broth, High Salt Formula, Powder is a specialized medium for the growth and cultivation of bacteria, particularly those requiring high salt concentrations for optimal growth. This 10kg powder formulation is ideal for large-scale microbial culture. The high salt concentration provides a stable osmotic environment, making it suitable for growing halophilic (salt-loving) microorganisms. This medium is commonly used in molecular biology, microbiology, and biotechnology laboratories for experiments requiring salt-tolerant bacterial species. Luria Broth supports robust bacterial growth and is often used in routine culture and fermentation processes, as well as for genetic analysis and other research applications.
Catalog No. 50-006-60578 Supplier Alkali Scientific Supplier No. LUA002CP
Luria Broth, High Salt Formula, Powder is a specialized medium for the growth and cultivation of bacteria, particularly those requiring high salt concentrations for optimal growth. This 10kg powder formulation is ideal for large-scale microbial culture. The high salt concentration provides a stable osmotic environment, making it suitable for growing halophilic (salt-loving) microorganisms. This medium is commonly used in molecular biology, microbiology, and biotechnology laboratories for experiments requiring salt-tolerant bacterial species. Luria Broth supports robust bacterial growth and is often used in routine culture and fermentation processes, as well as for genetic analysis and other research applications.