Learn More
Alkali Scientific Luria Broth, High Salt Formula, Powder [Miller's LB Broth], 2 Kg
SDP
Catalog No. 5000661451
Encompass_Preferred
Luria Broth, High Salt Formula, Powder (2kg) is a specialized microbiological medium designed to provide optimal conditions for growing salt-tolerant microorganisms. This high-salt version of Luria Broth is ideal for halophilic bacteria that thrive in high osmotic conditions. The powder form allows for easy preparation of liquid cultures by dissolving it in distilled water, providing a consistent environment for bacterial growth. It is frequently used in molecular biology and microbial research, particularly in applications requiring salt-stressed or halophilic microorganisms. This formulation is also utilized in fermentation and genetic studies that demand a controlled ionic strength environment.
Catalog No. 50-006-61451 Supplier Alkali Scientific Supplier No. LUA003CP
Luria Broth, High Salt Formula, Powder (2kg) is a specialized microbiological medium designed to provide optimal conditions for growing salt-tolerant microorganisms. This high-salt version of Luria Broth is ideal for halophilic bacteria that thrive in high osmotic conditions. The powder form allows for easy preparation of liquid cultures by dissolving it in distilled water, providing a consistent environment for bacterial growth. It is frequently used in molecular biology and microbial research, particularly in applications requiring salt-stressed or halophilic microorganisms. This formulation is also utilized in fermentation and genetic studies that demand a controlled ionic strength environment.