Learn More
Alkali Scientific Luria Broth, High Salt Formula, Powder [Miller's LB Broth], 500 G
SDP
Supplier:  Alkali Scientific LUA004CP
Encompass_Preferred
Luria Broth, High Salt Formula, Powder (500g) is a versatile medium used for cultivating salt-tolerant microorganisms, especially halophilic species. The high salt content ensures optimal growth conditions for bacteria that thrive in saline environments, making it ideal for microbiological research and industrial applications. This powder form allows for easy preparation of large or small batches, simply by dissolving the powder in water. Luria Broth is used in a variety of microbial culture applications, including genetic research, fermentation processes, and laboratory studies requiring high ionic strength for bacterial growth and maintenance.
Catalog No. 50-006-61642