Learn More
Alkali Scientific Luria Broth [Miller's LB] 1 Gram Buffered Capsules, 2.5 Kg
SDP
Catalog No. 5000659574
Encompass_Preferred
Luria Broth [Miller's LB] 1 Gram Buffered Capsules, 2.5 Kg is a bulk microbial culture medium designed for high-volume lab applications. The buffered formulation ensures that the pH remains stable throughout incubation, which is essential for supporting a wide range of microbial growth. The 2.5 Kg supply is ideal for laboratories that perform large-scale bacterial cultures or molecular biology experiments. Luria Broth is often used for the growth of E. coli and other microorganisms for DNA cloning, protein expression, and bacterial transformation. The capsules are easy to use, as they can be dissolved in water to create a consistent media solution.
Catalog No. 50-006-59574 Supplier Alkali Scientific Supplier No. IAB002CP
Luria Broth [Miller's LB] 1 Gram Buffered Capsules, 2.5 Kg is a bulk microbial culture medium designed for high-volume lab applications. The buffered formulation ensures that the pH remains stable throughout incubation, which is essential for supporting a wide range of microbial growth. The 2.5 Kg supply is ideal for laboratories that perform large-scale bacterial cultures or molecular biology experiments. Luria Broth is often used for the growth of E. coli and other microorganisms for DNA cloning, protein expression, and bacterial transformation. The capsules are easy to use, as they can be dissolved in water to create a consistent media solution.