Learn More
Alkali Scientific Luria Broth [Miller's LB] 1 Gram Buffered Capsules, 5 Kg
SDP
Catalog No. 5000659587
Encompass_Preferred
Luria Broth [Miller's LB] 1 Gram Buffered Capsules, 5 Kg is a large-volume culture medium used for the growth of bacteria, yeast, and other microorganisms in molecular biology applications. The buffered formulation ensures that the pH remains stable throughout incubation, which is vital for achieving consistent microbial growth. The 5 Kg packaging is perfect for high-throughput labs or those requiring large amounts of media for large-scale culturing or protein production. Luria Broth is commonly used for applications such as bacterial transformation, plasmid DNA preparation, and protein expression. The product is available in convenient buffered capsules, which dissolve quickly in water.
Catalog No. 50-006-59587 Supplier Alkali Scientific Supplier No. IAB004CP
Luria Broth [Miller's LB] 1 Gram Buffered Capsules, 5 Kg is a large-volume culture medium used for the growth of bacteria, yeast, and other microorganisms in molecular biology applications. The buffered formulation ensures that the pH remains stable throughout incubation, which is vital for achieving consistent microbial growth. The 5 Kg packaging is perfect for high-throughput labs or those requiring large amounts of media for large-scale culturing or protein production. Luria Broth is commonly used for applications such as bacterial transformation, plasmid DNA preparation, and protein expression. The product is available in convenient buffered capsules, which dissolve quickly in water.