Learn More
Alkali Scientific Luria Broth [Miller's LB] 1 Liter Buffered Capsules, 200 Capsules of 25 Grams, 5 Kg
SDP
Catalog No. 5000660615
Encompass_Preferred
Luria Broth [Miller's LB] 1 Liter Buffered Capsules, 200 Capsules of 25 Grams, 5 Kg is a large-volume culture medium designed for high-throughput laboratories or those requiring a large supply of growth media. The buffered formula ensures pH stability throughout the incubation process, which is crucial for maintaining optimal growth conditions for microorganisms. The 5 Kg size is ideal for labs that frequently perform large-scale culturing, bacterial transformation, or protein expression. This medium is commonly used in molecular biology applications for growing E. coli and other bacterial strains. The capsules are easily dissolved in water, making media preparation fast and convenient.
Catalog No. 50-006-60615 Supplier Alkali Scientific Supplier No. ABR003CP
Luria Broth [Miller's LB] 1 Liter Buffered Capsules, 200 Capsules of 25 Grams, 5 Kg is a large-volume culture medium designed for high-throughput laboratories or those requiring a large supply of growth media. The buffered formula ensures pH stability throughout the incubation process, which is crucial for maintaining optimal growth conditions for microorganisms. The 5 Kg size is ideal for labs that frequently perform large-scale culturing, bacterial transformation, or protein expression. This medium is commonly used in molecular biology applications for growing E. coli and other bacterial strains. The capsules are easily dissolved in water, making media preparation fast and convenient.