Learn More
Research Products International Corp Luria Broth Millers Mod 25KG
SDP
Catalog No. 502230462
Encompass_Preferred
For propagation and maintenace of E.Coli. Low Salt Formula. Molecular Biology Grade.
Dissolve 15.5 grams in 1 liter distilled water and adjust the pH to 7.2. Sterilize by autoclaving for 15 minutes.
Ingredients:
Casein Digest Peptone: 10 g/L
Yeast Extract: 5 g/L
Sodium Chloride: 0.5 g/L
Catalog No. 50-223-0462 Supplier Research Products International Corp Supplier No. L2408025000.0
For propagation and maintenace of E.Coli. Low Salt Formula. Molecular Biology Grade.
Dissolve 15.5 grams in 1 liter distilled water and adjust the pH to 7.2. Sterilize by autoclaving for 15 minutes.
Ingredients:
Casein Digest Peptone: 10 g/L
Yeast Extract: 5 g/L
Sodium Chloride: 0.5 g/L