Learn More
Medchemexpress LLC HY-U00117 5mg Medchemexpress, Lusaperidone CAS:214548-46-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYU001175MG
Encompass_Preferred
Medchemexpress, HY-U00117 5mg Lusaperidone CAS:214548-46-6 Lusaperidone (R107474) is an α2 adrenergic receptor antagonist with Kis of 0.13 and 0.15 nM for α2A and α2C, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0624