Learn More
Medchemexpress LLC Luseogliflozin hydrate | 1152425-66-5 | 99.2% | 452.56 | C23H32O7S | 10 MG
SDP
Catalog No. 5000242218
Encompass_Preferred
Luseogliflozin hydrate is the hydrated form of luseogliflozin, a selective and potent second-generation sodium-glucose cotransporter 2 (SGLT2) inhibitor used as a research reagent for studies of glucose transport and diabetes-related mechanisms. Supplied as an analytical-grade solid with high purity, it is intended for laboratory research use only.
  • High purity (99.2%) for reproducible results.
  • Molecular weight 452.56 and formula C23H32O7S.
  • Solid form suitable for accurate weighing and formulation.
  • Available in small package sizes for research-scale experiments.
  • Useful for SGLT2 inhibition and glucose transport studies.
  • Identified by CAS 1152425-66-5 for unambiguous referencing.
Catalog No. 50-002-42218 Supplier Medchemexpress LLC Supplier No. HY10449A10MG
Luseogliflozin hydrate is the hydrated form of luseogliflozin, a selective and potent second-generation sodium-glucose cotransporter 2 (SGLT2) inhibitor used as a research reagent for studies of glucose transport and diabetes-related mechanisms. Supplied as an analytical-grade solid with high purity, it is intended for laboratory research use only.
  • High purity (99.2%) for reproducible results.
  • Molecular weight 452.56 and formula C23H32O7S.
  • Solid form suitable for accurate weighing and formulation.
  • Available in small package sizes for research-scale experiments.
  • Useful for SGLT2 inhibition and glucose transport studies.
  • Identified by CAS 1152425-66-5 for unambiguous referencing.