Learn More
Medchemexpress LLC Luseogliflozin hydrate | 1152425-66-5 | 99.2% | 452.56 g/mol | C23H32O7S | 1 MG
SDP
Catalog No. 5000243200
Encompass_Preferred
Luseogliflozin hydrate is the hydrate form of luseogliflozin, a selective and potent second-generation sodium-glucose cotransporter 2 (SGLT2) inhibitor used in preclinical research into glucose transport and type 2 diabetes. Supplied in mg-scale quantities for laboratory use, the compound is well characterized for molecular formula, molecular weight, and analytical purity.
  • Selective SGLT2 inhibition for diabetes research.
  • High purity suitable for analytical and in vitro studies.
  • Available in multiple mg-scale pack sizes for flexible dosing.
  • Well characterized molecular formula and molecular weight.
Catalog No. 50-002-43200 Supplier Medchemexpress LLC Supplier No. HY10449A1MG
Luseogliflozin hydrate is the hydrate form of luseogliflozin, a selective and potent second-generation sodium-glucose cotransporter 2 (SGLT2) inhibitor used in preclinical research into glucose transport and type 2 diabetes. Supplied in mg-scale quantities for laboratory use, the compound is well characterized for molecular formula, molecular weight, and analytical purity.
  • Selective SGLT2 inhibition for diabetes research.
  • High purity suitable for analytical and in vitro studies.
  • Available in multiple mg-scale pack sizes for flexible dosing.
  • Well characterized molecular formula and molecular weight.