Learn More
Medchemexpress LLC Luseogliflozin hydrate | 1152425-66-5 | 99.2% | 452.56 g/mol | C23H32O7S | 25 MG
SDP
Catalog No. 5000242614
Encompass_Preferred
Luseogliflozin hydrate is a research-grade sodium-glucose cotransporter 2 (SGLT2) inhibitor provided as the hydrate form. It is a potent and selective compound used in preclinical studies of glucose homeostasis and type 2 diabetes mellitus. The material is supplied as a high-purity solid suitable for in vitro and in vivo research applications.
  • Potent SGLT2 inhibition (IC50 ≈ 2.26 nM).
  • High purity (99.2%) suitable for research use.
  • Hydrate form for consistent physicochemical properties.
  • Molecular weight 452.56 g/mol; formula C23H32O7S.
  • White to off-white solid, typically supplied in 25 MG quantities.
  • Suitable for in vitro and in vivo studies of glucose transport and metabolism.
Catalog No. 50-002-42614 Supplier Medchemexpress LLC Supplier No. HY10449A25MG
Luseogliflozin hydrate is a research-grade sodium-glucose cotransporter 2 (SGLT2) inhibitor provided as the hydrate form. It is a potent and selective compound used in preclinical studies of glucose homeostasis and type 2 diabetes mellitus. The material is supplied as a high-purity solid suitable for in vitro and in vivo research applications.
  • Potent SGLT2 inhibition (IC50 ≈ 2.26 nM).
  • High purity (99.2%) suitable for research use.
  • Hydrate form for consistent physicochemical properties.
  • Molecular weight 452.56 g/mol; formula C23H32O7S.
  • White to off-white solid, typically supplied in 25 MG quantities.
  • Suitable for in vitro and in vivo studies of glucose transport and metabolism.