Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Luseogliflozin hydrate | 1152425-66-5 | MFCD34574461 | 99.2% | 452.56 | C23H32O7S | 5 MG
SDP

Catalog No. 5000243482
Encompass_Preferred

Luseogliflozin hydrate is the hydrated form of luseogliflozin, a selective sodium-glucose cotransporter 2 (SGLT2) inhibitor used in preclinical and in vitro research on glucose transport and type 2 diabetes. The compound is supplied for laboratory research with accompanying analytical and safety documentation.

  • Purity 99.2%.
  • Molecular weight 452.56 g/mol.
  • CAS number 1152425-66-5.
  • Available as a solid powder in multiple pack sizes including 5 MG.
  • Analytical data available: COA, HNMR, LCMS, SDS.
  • Intended for preclinical and in vitro research use only.

Catalog No. 50-002-43482 Supplier Medchemexpress LLC Supplier No. HY10449A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Luseogliflozin hydrate is the hydrated form of luseogliflozin, a selective sodium-glucose cotransporter 2 (SGLT2) inhibitor used in preclinical and in vitro research on glucose transport and type 2 diabetes. The compound is supplied for laboratory research with accompanying analytical and safety documentation.

  • Purity 99.2%.
  • Molecular weight 452.56 g/mol.
  • CAS number 1152425-66-5.
  • Available as a solid powder in multiple pack sizes including 5 MG.
  • Analytical data available: COA, HNMR, LCMS, SDS.
  • Intended for preclinical and in vitro research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.