Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Luseogliflozin hydrate | 1152425-66-5 | MFCD34574461 | 99.2% | 452.56 | C23H32O7S | 50 MG
SDP

Catalog No. 5000243470
Encompass_Preferred

Luseogliflozin hydrate is a research-grade sodium-glucose cotransporter 2 (SGLT2) inhibitor supplied as a white to off-white solid for biochemical and pharmacological studies. It is intended for in vitro and in vivo research on glucose transport and metabolic disease and should be handled and stored under recommended conditions.

  • High purity (99.2%) suitable for research applications.
  • Molecular weight 452.56 g/mol for accurate dosing and calculations.
  • White to off-white solid form for ease of handling.
  • Storage: sealed at -20°C; in solvent store at -80°C for long-term stability.
  • Used in studies of SGLT2-mediated glucose transport and metabolic disease models.

Catalog No. 50-002-43470 Supplier Medchemexpress LLC Supplier No. HY10449A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Luseogliflozin hydrate is a research-grade sodium-glucose cotransporter 2 (SGLT2) inhibitor supplied as a white to off-white solid for biochemical and pharmacological studies. It is intended for in vitro and in vivo research on glucose transport and metabolic disease and should be handled and stored under recommended conditions.

  • High purity (99.2%) suitable for research applications.
  • Molecular weight 452.56 g/mol for accurate dosing and calculations.
  • White to off-white solid form for ease of handling.
  • Storage: sealed at -20°C; in solvent store at -80°C for long-term stability.
  • Used in studies of SGLT2-mediated glucose transport and metabolic disease models.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.