Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Luseogliflozin hydrate | 1152425-66-5 | MFCD34574461 | 99.2% | 452.56 g/mol | C23H32O7S | 100 MG
SDP

Catalog No. 5000243199
Encompass_Preferred

Luseogliflozin hydrate is a selective, potent, orally active sodium-glucose cotransporter 2 (SGLT2) inhibitor supplied as a high-purity solid for research use. It is intended for biochemical and pharmacological studies of glucose transport and metabolic disease models such as type 2 diabetes.

  • High purity (99.2%).
  • Molecular weight 452.56 g/mol.
  • CAS number 1152425-66-5.
  • White to off-white solid powder.
  • Intended for SGLT2-related biochemical and pharmacological research.
  • Available in small research pack sizes such as 100 mg.

Catalog No. 50-002-43199 Supplier Medchemexpress LLC Supplier No. HY10449A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Luseogliflozin hydrate is a selective, potent, orally active sodium-glucose cotransporter 2 (SGLT2) inhibitor supplied as a high-purity solid for research use. It is intended for biochemical and pharmacological studies of glucose transport and metabolic disease models such as type 2 diabetes.

  • High purity (99.2%).
  • Molecular weight 452.56 g/mol.
  • CAS number 1152425-66-5.
  • White to off-white solid powder.
  • Intended for SGLT2-related biochemical and pharmacological research.
  • Available in small research pack sizes such as 100 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.