Learn More
Medchemexpress LLC Luspatercept, 1mg
SDP
Supplier:  Medchemexpress LLC HYP997201MG
Encompass_Preferred
Luspatercept (ACE-536) is a recombinant modified ActRIIB fusion protein that binds with transforming growth factor β superfamily ligands. Luspatercept increases the erythrocyte numbers and promotes maturation of erythroid precursors. Luspatercept binds with GDF11 and inhibits Smad2/3 signaling. Luspatercept can be used for the research of anemia.
Catalog No. NC2822307