Learn More
Cayman Chemical Lusutrombopag 10mg
SDP
Catalog No. 502935437
Encompass_Preferred
A thrombopoietin receptor agonist; induces proliferation of Ba/F3 cells expressing human MPL (EC50 = 84 nM) but not of those expressing mouse Mpl; increases the number of circulating platelets in a transgenic mouse model expressing human MPL at 0.3-10 mg/kg per day and increases the number of megakaryocytes in the bone marrow at 10 mg/kg per day
Catalog No. 50-293-5437 Supplier Cayman Chemical Supplier No. 2653310 MG
A thrombopoietin receptor agonist; induces proliferation of Ba/F3 cells expressing human MPL (EC50 = 84 nM) but not of those expressing mouse Mpl; increases the number of circulating platelets in a transgenic mouse model expressing human MPL at 0.3-10 mg/kg per day and increases the number of megakaryocytes in the bone marrow at 10 mg/kg per day