Learn More
Medchemexpress LLC HY-111940 10mg Medchemexpress, LUT014 CAS:2274819-46-2 Purity:>98%
SDP
Catalog No. 501968988
Encompass_Preferred
Medchemexpress, HY-111940 10mg LUT014 CAS:2274819-46-2 LUT014 is a B-Raf inhibitor with an IC50 of 11.7 nM, and developed to reduce dose-limiting acneiform lesions associated EGFR Inhibitors treatment. Extracted from patent WO 2019026065A2 . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8988 Supplier Medchemexpress LLC Supplier No. HY11194010MG
Medchemexpress, HY-111940 10mg LUT014 CAS:2274819-46-2 LUT014 is a B-Raf inhibitor with an IC50 of 11.7 nM, and developed to reduce dose-limiting acneiform lesions associated EGFR Inhibitors treatment. Extracted from patent WO 2019026065A2 . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.