Learn More
Med Vet International Zoetis, Lutalyse, Highcon, Injection, 20mL
Catalog No. 502828323
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
LUTALYSE HighCon Injection (dinoprost tromthamine injection) is a high-concentration formula of the proven LUTALYSE Injection (dinoprost injection) used to help improve breeding efficiency in beef and dairy cattle. Like LUTALYSE, LUTALYSE HighCon is approved for use with FACTREL Injection (gonadorelin injection) to synchronize estrous cycles to allow for fixed- time artificial insemination (FTAI) in lacatin cows, for yielding higher pregnancy rates and improved calving intervals.
Catalog No. 50-282-8323 Supplier Med Vet International Supplier No. RXLUTY20
 Restricted Item
Add to Cart
Add to Cart
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
LUTALYSE HighCon Injection (dinoprost tromthamine injection) is a high-concentration formula of the proven LUTALYSE Injection (dinoprost injection) used to help improve breeding efficiency in beef and dairy cattle. Like LUTALYSE, LUTALYSE HighCon is approved for use with FACTREL Injection (gonadorelin injection) to synchronize estrous cycles to allow for fixed- time artificial insemination (FTAI) in lacatin cows, for yielding higher pregnancy rates and improved calving intervals.