Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lutein, ≥96% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111012895 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2895 1 mg
11-101-2896 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-2895 Supplier MilliporeSigma Supelco Supplier No. 071681MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lutein is a yellow dihydroxylated carotenoid which is found to be a common constituent in many dietary supplements.

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 127-40-2
Physical Form Powder
Linear Formula C40H56O2
MDL Number MFCD08435941
Synonym Xanthophyll; alpha-Carotene-3,3′-diol; beta, epsilon-carotene; beta, epsilon-carotene-3,3′-diol; Lutein
Type Analytical Standard
Quantity 1 mg
Formula Weight 568.87
Percent Purity ≥96% (HPLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.