Learn More
Cusabio Technology LLC Luteinizing hormone LH
Catalog No. 5000048769
Encompass_Preferred
This kit detects luteinizing hormone (LH) in human serum plasma and tissue homogenates using a sandwich ELISA technique with a measurable range of 1-50 mIU/ml and a sensitivity of 0 5 mIU/ml Designed for quantitative analysis the assay requires a sample volume of 50-100 L and employs a 450 nm absorbance detection method The optimized protocol ensures results within 1-5 hours of incubation making it suitable for clinical diagnostics or research settings requiring precise measurement of LH levels The kit s robust performance and standardized workflow support reliable detection of physiological or pathological LH variations across diverse biological samples
Catalog No. 50-000-48769 Supplier Cusabio Technology LLC Supplier No. CSBE12690H
This kit detects luteinizing hormone (LH) in human serum plasma and tissue homogenates using a sandwich ELISA technique with a measurable range of 1-50 mIU/ml and a sensitivity of 0 5 mIU/ml Designed for quantitative analysis the assay requires a sample volume of 50-100 L and employs a 450 nm absorbance detection method The optimized protocol ensures results within 1-5 hours of incubation making it suitable for clinical diagnostics or research settings requiring precise measurement of LH levels The kit s robust performance and standardized workflow support reliable detection of physiological or pathological LH variations across diverse biological samples