Learn More
Creative Biomart Buserelin
Supplier:  Creative Biomart THP0008
Encompass_Preferred
The product is a synthetic peptide analog of the luteinizing hormone-releasing hormone (LHRH) agonist, which stimulates the pituitary gland's gonadotrophin-releasing hormone receptor (GnRHR).
Catalog No. 50-208-4059