Learn More
CPC Scientific Ac-D-2-Nal-p-chloro-D-Phe-beta-(3-pyridyl)-D-Ala-Gly-Arg-Pro-D-Ala-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific LHRH021A
Encompass
This peptide showing high receptor binding affinity (IC50 = 7 nM) has been designed by truncation strategy from antide.SEQUENCE: Ac-D-2-Nal-p-chloro-D-Phe-beta-(3-pyridyl)-D-Ala-Gly-Arg-Pro-D-Ala-NH2 (trifluoroacetate salt)ONE-LETTER SEQUENCE: Ac-D-2-Nal-p-chloro-f-beta-(3-pyridyl)-AGRPa-NH2MOLECULAR FORMULA: C48H59Cl1N12O8MOLECULAR WEIGHT: 967.53STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [292141-31-2]SYNONYMS: RESEARCH AREA: HormonalREFERENCES:D.Yahalom et al., J. Med. Chem., 43, 2831 (2000)SKU(s): LHRH-021A, LHRH-021B
Catalog No. 50-210-9673