Learn More
Apexbio Technology LLC Luteinizing hormone releasing hormone human acetate salt (LHRH) 33515-09-2 1mg
SDP
Catalog No. 5000095549
Encompass_Preferred
Luteinizing hormone releasing hormone human acetate salt (LHRH) is a neuropeptide hormone targeting anterior pituitary signaling pathways It is designed to stimulate the anterior pituitary gland thereby promoting secretion of follicle-stimulating hormone (FSH) and luteinizing hormone (LH) LHRH exerts its biological activity primarily by selectively activating matrix metalloproteinase-2 (MMP-2) and matrix metalloproteinase-9 (MMP-9) key enzymes involved in extracellular matrix remodeling In studies with human endometrial cancer cell lines and decidual stromal cells LHRH mediates induction of MMP-9 expression and activation Based on these pharmacological properties LHRH holds research potential in reproductive biology embryo implantation tumor cell invasiveness and metastasis studies
Catalog No. 50-000-95549 Supplier Apexbio Technology LLC Supplier No. A11471
Add to Cart
Add to Cart
Luteinizing hormone releasing hormone human acetate salt (LHRH) is a neuropeptide hormone targeting anterior pituitary signaling pathways It is designed to stimulate the anterior pituitary gland thereby promoting secretion of follicle-stimulating hormone (FSH) and luteinizing hormone (LH) LHRH exerts its biological activity primarily by selectively activating matrix metalloproteinase-2 (MMP-2) and matrix metalloproteinase-9 (MMP-9) key enzymes involved in extracellular matrix remodeling In studies with human endometrial cancer cell lines and decidual stromal cells LHRH mediates induction of MMP-9 expression and activation Based on these pharmacological properties LHRH holds research potential in reproductive biology embryo implantation tumor cell invasiveness and metastasis studies