Learn More
Alpco Ltd Luteinizing Hormone ELISA (LH ELISA) (Urine)
SDP
Supplier:  Alpco Ltd 20LHUHUE01
Encompass_Preferred
The LH (Urine) ELISA is an enzyme immunoassay for the quantitative in vitro diagnostic measurement of LH in urine. This test is used to detect the midcycle LH surge in urine, which is an aid in predicting the time of ovulation. FDA Registered. For In Vitro Diagnostic Use.
Catalog No. 50-305-111