Learn More
Apexbio Technology LLC Luteolin, 20mg. Cas: 491-70-3 MFCD: MFCD00017309. Antioxidant and free radical scavenger
SDP
Supplier:  Apexbio Technology LLC N183120
Encompass_Preferred
Anti-inflammatory, antioxidant and free radical scavenger. Inhibits LPS-induced TNF-, IL-6 and inducible nitric oxide production and blocks NF-B and AP-1 activation. Also inhibits TNF- -induced COX-2 expression. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5644
 Restricted Item