Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Luteolin-3′,7-diglucoside | 52187-80-1 | 1 MG
SDP

Catalog No. 5000417400
Encompass_Preferred

Luteolin-3′,7-diglucoside is a glycoside isolated from *R. luteolin* that significantly inhibits cataracts induced in ovine lenses and inhibits the activity of *Ureaplasma urealyticum* ATCC and clinical strains. It works by counteracting IL1-induced tissue factor expression in monocytes, thus inhibiting procoagulant activity. This compound also demonstrates a significant ability to inhibit cataracts in ovine lenses when incubated in hypotonic hepes buffered solution.

  • Molecular weight: 610.52
  • Formula: C₂₇H₃₀O₁₆
  • Appearance: Solid
  • Color: Light yellow to yellow
  • Structure classification: Flavonoids, isoflavones
  • Initial source: Plants, Solanaceae, *Alkekengi officinarum Moench*
  • Inhibits procoagulant activity in monocytes
  • Significantly inhibits cataracts induced in ovine lenses
  • Inhibits *Ureaplasma urealyticum* activity

Catalog No. 50-004-17400 Supplier Medchemexpress LLC Supplier No. HYN83181MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Luteolin-3′,7-diglucoside is a glycoside isolated from *R. luteolin* that significantly inhibits cataracts induced in ovine lenses and inhibits the activity of *Ureaplasma urealyticum* ATCC and clinical strains. It works by counteracting IL1-induced tissue factor expression in monocytes, thus inhibiting procoagulant activity. This compound also demonstrates a significant ability to inhibit cataracts in ovine lenses when incubated in hypotonic hepes buffered solution.

  • Molecular weight: 610.52
  • Formula: C₂₇H₃₀O₁₆
  • Appearance: Solid
  • Color: Light yellow to yellow
  • Structure classification: Flavonoids, isoflavones
  • Initial source: Plants, Solanaceae, *Alkekengi officinarum Moench*
  • Inhibits procoagulant activity in monocytes
  • Significantly inhibits cataracts induced in ovine lenses
  • Inhibits *Ureaplasma urealyticum* activity

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.