Learn More
Medchemexpress LLC Luteolin-3',7-diglucoside | 52187-80-1 | 5 MG
SDP
Catalog No. 5000419181
Encompass_Preferred
Luteolin-3′,7-diglucoside is a glycoside that can be isolated from *R. luteolin*. It significantly inhibits cataracts induced in ovine lenses and also inhibits the activity of *Ureaplasma urealyticum* ATCC and clinical strains.
  • Glycoside isolated from *R. luteolin*.
  • Significantly inhibits cataracts induced in ovine lenses.
  • Inhibits activity of *Ureaplasma urealyticum* ATCC and clinical strains.
  • High purity of 97.0%.
  • Appears as a solid, light yellow to yellow.
Catalog No. 50-004-19181 Supplier Medchemexpress LLC Supplier No. HYN83185MG
Luteolin-3′,7-diglucoside is a glycoside that can be isolated from *R. luteolin*. It significantly inhibits cataracts induced in ovine lenses and also inhibits the activity of *Ureaplasma urealyticum* ATCC and clinical strains.
  • Glycoside isolated from *R. luteolin*.
  • Significantly inhibits cataracts induced in ovine lenses.
  • Inhibits activity of *Ureaplasma urealyticum* ATCC and clinical strains.
  • High purity of 97.0%.
  • Appears as a solid, light yellow to yellow.