Learn More
Medchemexpress LLC Luteolin-3-O-beta-D-glucuronide | 53527-42-7 | 98.9% | 462.36 g·mol⁻¹ | C21H18O12 | 10 MG
SDP
Catalog No. 5000289579
Encompass_Preferred
Luteolin-3-O-beta-D-glucuronide is a flavonoid metabolite of luteolin used as a research reagent and analytical standard in biochemical and pharmacological studies. The compound is supplied as a high-purity solid for use in metabolism, enzymology, and analytical method development.
  • High purity (~98.9%) suitable for analytical and biological assays.
  • Molecular formula C21H18O12 and molecular weight 462.36 g·mol⁻¹.
  • Identified by CAS number 53527-42-7 for unambiguous chemical tracking.
  • Available in small milligram quantities for assay development and reference use.
  • Reported as a luteolin metabolite present in plant sources such as Rosmarinus officinalis.
Catalog No. 50-002-89579 Supplier Medchemexpress LLC Supplier No. HYN409910MG
Luteolin-3-O-beta-D-glucuronide is a flavonoid metabolite of luteolin used as a research reagent and analytical standard in biochemical and pharmacological studies. The compound is supplied as a high-purity solid for use in metabolism, enzymology, and analytical method development.
  • High purity (~98.9%) suitable for analytical and biological assays.
  • Molecular formula C21H18O12 and molecular weight 462.36 g·mol⁻¹.
  • Identified by CAS number 53527-42-7 for unambiguous chemical tracking.
  • Available in small milligram quantities for assay development and reference use.
  • Reported as a luteolin metabolite present in plant sources such as Rosmarinus officinalis.