Learn More
AdipoGen Luteolin
SDP
Catalog No. 502560602
Encompass_Preferred
Chemical. CAS 491-70-3. Formula C15H10O6. MW 286.24. Luteolin is a hydroxylated flavone derivative that is a strong antioxidant and radical scavenger. It has multiple activities, such as anticancer, antimetastatic, anti-adipogenic, anti-inflammatory and neuroprotective. Luteolin has been described as a fatty acid synthase FASN, 15-lipoxygenase, alpha-glucosidase, PPAR-gamma, topoisomerase I, PI3K/Akt, RSK1/RSK2 and HDAC inhibitor.
Catalog No. 50-256-0602 Supplier AdipoGen Supplier No. CDXL0087M010
Chemical. CAS 491-70-3. Formula C15H10O6. MW 286.24. Luteolin is a hydroxylated flavone derivative that is a strong antioxidant and radical scavenger. It has multiple activities, such as anticancer, antimetastatic, anti-adipogenic, anti-inflammatory and neuroprotective. Luteolin has been described as a fatty acid synthase FASN, 15-lipoxygenase, alpha-glucosidase, PPAR-gamma, topoisomerase I, PI3K/Akt, RSK1/RSK2 and HDAC inhibitor.