Learn More
AdipoGen Luteolin
SDP
Catalog No. 502562662
Encompass_Preferred
Chemical. CAS 491-70-3. Formula C15H10O6. MW 286.2. Synthetic. Anticancer and antimetastatic. alpha-Glucosidase inhibitor. Topoisomerase I Inhibitor. Apoptosis inducer. Anti-inflammatory. 15-Lipoxygenase 15-LOX inhibitor. IL-6 production inhibitor. Fatty acid synthase FAS inhibitor. Antioxidant. Free radical scavenger. Antiadipogenic. PPAR-gamma inhibitor. Neuroprotective. Monoamine transporter activator. Inhibitor of phosphodiesterases 1-5 PDE1-5. PKCepsilon and Src kinase inhibitor. HDAC inhibitor. Autophagy modulator Phosphatidylinositol 3-kinase PI3K / Akt signaling inhibitor. p90 ribosomal S6 kinase RSK1/RSK2 kinase activity inhibitor.
Catalog No. 50-256-2662 Supplier AdipoGen Supplier No. AGCN20098M005
Chemical. CAS 491-70-3. Formula C15H10O6. MW 286.2. Synthetic. Anticancer and antimetastatic. alpha-Glucosidase inhibitor. Topoisomerase I Inhibitor. Apoptosis inducer. Anti-inflammatory. 15-Lipoxygenase 15-LOX inhibitor. IL-6 production inhibitor. Fatty acid synthase FAS inhibitor. Antioxidant. Free radical scavenger. Antiadipogenic. PPAR-gamma inhibitor. Neuroprotective. Monoamine transporter activator. Inhibitor of phosphodiesterases 1-5 PDE1-5. PKCepsilon and Src kinase inhibitor. HDAC inhibitor. Autophagy modulator Phosphatidylinositol 3-kinase PI3K / Akt signaling inhibitor. p90 ribosomal S6 kinase RSK1/RSK2 kinase activity inhibitor.