Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Luteolin-4'-O-glucoside | 6920-38-3 | 1 MG
SDP

Catalog No. 5000417399
Encompass_Preferred

Luteolin-4'-O-glucoside is an IL-5 inhibitor with an IC50 of 3.7 μM. It resists hyperuricemia and acute gouty arthritis activity and also exhibits anticancer activity. This compound has a molecular weight of 448.38, a chemical formula of C21H20O11, and appears as an off-white to yellow solid. It is classified as a flavonoid and phenol, and is initially sourced from plants such as Punicaceae Punica granatum L. This product is intended for research use only.

  • IL-5 inhibitor with an IC50 of 3.7 μM
  • Resists hyperuricemia and acute gouty arthritis activity
  • Exhibits anticancer activity
  • Molecular weight of 448.38
  • Chemical formula C21H20O11
  • Solid appearance, off-white to yellow color
  • 99.36% purity
  • Cytotoxic activity against NCI-H929, U266, and OPM2 cell lines
  • Derived from plants like Punicaceae Punica granatum L.
  • For research use only

Catalog No. 50-004-17399 Supplier Medchemexpress LLC Supplier No. HYN83091MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Luteolin-4'-O-glucoside is an IL-5 inhibitor with an IC50 of 3.7 μM. It resists hyperuricemia and acute gouty arthritis activity and also exhibits anticancer activity. This compound has a molecular weight of 448.38, a chemical formula of C21H20O11, and appears as an off-white to yellow solid. It is classified as a flavonoid and phenol, and is initially sourced from plants such as Punicaceae Punica granatum L. This product is intended for research use only.

  • IL-5 inhibitor with an IC50 of 3.7 μM
  • Resists hyperuricemia and acute gouty arthritis activity
  • Exhibits anticancer activity
  • Molecular weight of 448.38
  • Chemical formula C21H20O11
  • Solid appearance, off-white to yellow color
  • 99.36% purity
  • Cytotoxic activity against NCI-H929, U266, and OPM2 cell lines
  • Derived from plants like Punicaceae Punica granatum L.
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.