Learn More
Medchemexpress LLC Luteolin | 491-70-3 | 99.51% | 286.24 | 10 MG
SDP
Catalog No. 5000365263
Encompass_Preferred
Luteolin, also known as Luteoline or Luteolol, is a flavonoid compound and a potent Nrf2 inhibitor. It possesses anti-inflammatory and anti-cancer properties. This product is for research use only and is not sold to patients.
  • Induces apoptosis and cell cycle arrest
  • Inhibits metastasis and angiogenesis in various cancer cell lines
  • Suitable for research applications
Catalog No. 50-003-65263 Supplier Medchemexpress LLC Supplier No. 10MGHYN0162
Luteolin, also known as Luteoline or Luteolol, is a flavonoid compound and a potent Nrf2 inhibitor. It possesses anti-inflammatory and anti-cancer properties. This product is for research use only and is not sold to patients.
  • Induces apoptosis and cell cycle arrest
  • Inhibits metastasis and angiogenesis in various cancer cell lines
  • Suitable for research applications