Learn More
Medchemexpress LLC Luteolin | 491-70-3 | MFCD00006839 | 99.51% | 286.24 | 50 MG
SDP
Catalog No. 5000427596
Encompass_Preferred
Luteolin, a flavanoid compound, is a potent Nrf2 inhibitor. It exhibits anti-inflammatory and anti-cancer properties, including the induction of apoptosis and cell cycle arrest, and the inhibition of metastasis and angiogenesis, in several cancer cell lines, such as human non-small lung cancer cells.
  • Acts as a potent Nrf2 inhibitor
  • Exhibits anti-inflammatory and anti-cancer properties
  • Induces apoptosis and cell cycle arrest
  • Inhibits metastasis and angiogenesis
  • Effective in various cancer cell lines
  • Utilized for assessing cell viability
  • Applied in immunofluorescence analysis
  • Used for hematoxylin and eosin staining
  • Investigated for effects on protein levels via western blotting
Catalog No. 50-004-27596 Supplier Medchemexpress LLC Supplier No. HYN0162R50MG
Luteolin, a flavanoid compound, is a potent Nrf2 inhibitor. It exhibits anti-inflammatory and anti-cancer properties, including the induction of apoptosis and cell cycle arrest, and the inhibition of metastasis and angiogenesis, in several cancer cell lines, such as human non-small lung cancer cells.
  • Acts as a potent Nrf2 inhibitor
  • Exhibits anti-inflammatory and anti-cancer properties
  • Induces apoptosis and cell cycle arrest
  • Inhibits metastasis and angiogenesis
  • Effective in various cancer cell lines
  • Utilized for assessing cell viability
  • Applied in immunofluorescence analysis
  • Used for hematoxylin and eosin staining
  • Investigated for effects on protein levels via western blotting