Learn More
Medchemexpress LLC Luteolin 5-O-glucoside | 20344-46-1 | 448.38 | 5 MG
SDP
Catalog No. 5000415416
Encompass_Preferred
Luteolin 5-O-glucoside is a major flavonoid derived from Cirsium maackii with notable anti-inflammatory activity. It is known to inhibit LPS-induced NO production and t-BHP-induced ROS generation. This compound also suppresses the expression of iNOS and COX-2 in macrophages.
  • Exhibits anti-inflammatory activity
  • Inhibits LPS-induced NO production
  • Inhibits t-BHP-induced ROS generation
  • Suppresses the expression of iNOS and COX-2 in macrophages
  • Effective at non-toxic concentrations in RAW 264.7 cells
Catalog No. 50-004-15416 Supplier Medchemexpress LLC Supplier No. HYN20085MG
Luteolin 5-O-glucoside is a major flavonoid derived from Cirsium maackii with notable anti-inflammatory activity. It is known to inhibit LPS-induced NO production and t-BHP-induced ROS generation. This compound also suppresses the expression of iNOS and COX-2 in macrophages.
  • Exhibits anti-inflammatory activity
  • Inhibits LPS-induced NO production
  • Inhibits t-BHP-induced ROS generation
  • Suppresses the expression of iNOS and COX-2 in macrophages
  • Effective at non-toxic concentrations in RAW 264.7 cells