Learn More
Medchemexpress LLC Luteolin 5-O-glucoside | 20344-46-1 | MFCD11656133 | ≥98.0% | 448.38 g·mol⁻¹ | C21H20O11 | 10 MG
SDP
Catalog No. 5000295720
Encompass_Preferred
Luteolin 5-O-glucoside is a flavonoid O-glycoside used in research to study anti-inflammatory and antioxidant mechanisms. It inhibits LPS-induced nitric oxide production and t-BHP-induced reactive oxygen species generation, and is supplied as a high-purity solid for biochemical and cell-based assays.
  • Flavonoid glycoside with anti-inflammatory activity.
  • Inhibits LPS-induced NO production and t-BHP-induced ROS generation.
  • High purity suitable for biochemical and cell-based assays.
  • Solid, light yellow to yellow appearance for easy handling.
  • Packaged as a small research vial (10 mg) for screening and exploratory studies.
Catalog No. 50-002-95720 Supplier Medchemexpress LLC Supplier No. HYN200810MG
Luteolin 5-O-glucoside is a flavonoid O-glycoside used in research to study anti-inflammatory and antioxidant mechanisms. It inhibits LPS-induced nitric oxide production and t-BHP-induced reactive oxygen species generation, and is supplied as a high-purity solid for biochemical and cell-based assays.
  • Flavonoid glycoside with anti-inflammatory activity.
  • Inhibits LPS-induced NO production and t-BHP-induced ROS generation.
  • High purity suitable for biochemical and cell-based assays.
  • Solid, light yellow to yellow appearance for easy handling.
  • Packaged as a small research vial (10 mg) for screening and exploratory studies.