Learn More
Medchemexpress LLC HY-N0162 500mg , Luteolin CAS:491-70-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN0162500MG
Encompass_Preferred
HY-N0162 500mg Luteolin CAS: 491-70-3 Luteolin (Luteoline), a flavanoid compound, is a potent Nrf2 inhibitor. Luteolin has anti-inflammatory, anti-cancer properties, including the induction of apoptosis and cell cycle arrest, and the inhibition of metastasis and angiogenesis, in several cancer cell lines, including human non-small lung cancer cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1170