Learn More
TARGETMOL CHEMICALS INC Luteolin 500MG
SDP
Catalog No. 502880765
Encompass_Preferred
Also available in 1 g, 1 mL, 10 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Luteolin (Luteolol) belongs to the flavonoid group of natural products and is a Nrf2 inhibitor, PDE inhibitor. Luteolin has a wide range of biological activities including antitumor, antioxidant, anti-inflammatory, antimicrobial, antiviral, antiallergic, and procoagulant. Purity 97.39%
Catalog No. 50-288-0765 Supplier TARGETMOL CHEMICALS INC Supplier No. T1027500MG
Also available in 1 g, 1 mL, 10 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Luteolin (Luteolol) belongs to the flavonoid group of natural products and is a Nrf2 inhibitor, PDE inhibitor. Luteolin has a wide range of biological activities including antitumor, antioxidant, anti-inflammatory, antimicrobial, antiviral, antiallergic, and procoagulant. Purity 97.39%