Learn More
Selleck Chemical LLC Luteolin S2320-10mg
SDP
Catalog No. 503109556
Encompass_Preferred
Luteolin (Luteoline Luteolol Digitoflavone) is a flavonoid found in Terminalia chebula which is a non-selective phisphodiesterase PDE inhibitor for PDE1-5 with Ki of 15 0 M 6 4 M 13 9 M 11 1 M and 9 5 M respectively
Catalog No. 50-310-9556 Supplier Selleck Chemical LLC Supplier No. S232010MG
Luteolin (Luteoline Luteolol Digitoflavone) is a flavonoid found in Terminalia chebula which is a non-selective phisphodiesterase PDE inhibitor for PDE1-5 with Ki of 15 0 M 6 4 M 13 9 M 11 1 M and 9 5 M respectively