Learn More
Selleck Chemical LLC Luteolin S2320-1g
SDP
Catalog No. 503111778
Encompass_Preferred
Luteolin (Luteoline Luteolol Digitoflavone) is a flavonoid found in Terminalia chebula which is a non-selective phisphodiesterase PDE inhibitor for PDE1-5 with Ki of 15 0 M 6 4 M 13 9 M 11 1 M and 9 5 M respectively
Catalog No. 50-311-1778 Supplier Selleck Chemical LLC Supplier No. S23201G
Luteolin (Luteoline Luteolol Digitoflavone) is a flavonoid found in Terminalia chebula which is a non-selective phisphodiesterase PDE inhibitor for PDE1-5 with Ki of 15 0 M 6 4 M 13 9 M 11 1 M and 9 5 M respectively