Learn More
TARGETMOL CHEMICALS INC Luteolin 7-diglucuronide 5MG
SDP
Catalog No. 502875847
Encompass_Preferred
Also available in 1 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Luteolin 7-diglucuronide, the major flavonoid isolated from Verbena officinalis and Aloysia triphylla. Purity 98.44%
Catalog No. 50-287-5847 Supplier TARGETMOL CHEMICALS INC Supplier No. T137615MG
Also available in 1 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Luteolin 7-diglucuronide, the major flavonoid isolated from Verbena officinalis and Aloysia triphylla. Purity 98.44%