Learn More
Cayman Chemical LuteolIn 7-O-GlucuronIde 1mg
SDP
Catalog No. 502946341
Encompass_Preferred
A flavonoid with antioxidant, enzyme inhibitory, and anti-inflammatory properties; scavenges DPPH radicals in a cell-free assay (IC50 = 80.6 μM); inhibits porcine α-amylase and yeast α-glucosidase in vitro (IC50s = 61.5 and 14.7 μM, respectively); inhibits rat aldose reductase in vitro (IC50 = 0.7 μM) and sorbitol accumulation in isolated rat lens at 5 μg/ml; inhibits LPS-induced cytokine release from RAW 264.7 macrophages in a dose-dependent manner
Catalog No. 50-294-6341 Supplier Cayman Chemical Supplier No. 331881 MG
A flavonoid with antioxidant, enzyme inhibitory, and anti-inflammatory properties; scavenges DPPH radicals in a cell-free assay (IC50 = 80.6 μM); inhibits porcine α-amylase and yeast α-glucosidase in vitro (IC50s = 61.5 and 14.7 μM, respectively); inhibits rat aldose reductase in vitro (IC50 = 0.7 μM) and sorbitol accumulation in isolated rat lens at 5 μg/ml; inhibits LPS-induced cytokine release from RAW 264.7 macrophages in a dose-dependent manner