Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Luteolin 7-O-glucuronide | 29741-10-4 | 98.7% | 462.36 | C21H18O12 | 10 MG
SDP

Catalog No. 5000295514
Encompass_Preferred

Luteolin 7-O-glucuronide is a flavone glycoside used as a research reagent to study matrix metalloproteinase (MMP) inhibition and inflammation-related pathways. It is supplied as a light-yellow solid and as a DMSO solution, and is provided at high purity for biochemical assays and analytical standards.

  • Cas number: 29741-10-4.
  • Molecular weight: 462.36.
  • Typical purity: 98.7% (batch level).
  • Physical forms: solid and 10 mM solution in DMSO.
  • Typical package sizes include 1 mg, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg.
  • Common applications: enzymatic inhibition assays and analytical standardization.

Catalog No. 50-002-95514 Supplier Medchemexpress LLC Supplier No. HYN146310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Luteolin 7-O-glucuronide is a flavone glycoside used as a research reagent to study matrix metalloproteinase (MMP) inhibition and inflammation-related pathways. It is supplied as a light-yellow solid and as a DMSO solution, and is provided at high purity for biochemical assays and analytical standards.

  • Cas number: 29741-10-4.
  • Molecular weight: 462.36.
  • Typical purity: 98.7% (batch level).
  • Physical forms: solid and 10 mM solution in DMSO.
  • Typical package sizes include 1 mg, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg.
  • Common applications: enzymatic inhibition assays and analytical standardization.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.