Learn More
Cayman Chemical Luteolin 7-O-Rutinoside, Purity: ≥95%, CAS: 20633-84-5, 10 mg
SDP
Catalog No. 502946528
Encompass_Preferred
A flavonoid glycoside with diverse biological activities; scavenges DPPH radicals in a cell-free assay from 1-50 µM; active against various bacteria (MICs = 100-200 µg/ml); active against C. albicans, S. cerevisiae, and C. lusitaniae (MICs = 100, 200, and 50 µg/ml, respectively); decreases hepatic and renal injury and increases survival in a mouse model of polyphosphate-induced lethal endotoxemia at 57.6 µg/animal
Catalog No. 50-294-6528 Supplier Cayman Chemical Supplier No. 3403510 MG
A flavonoid glycoside with diverse biological activities; scavenges DPPH radicals in a cell-free assay from 1-50 µM; active against various bacteria (MICs = 100-200 µg/ml); active against C. albicans, S. cerevisiae, and C. lusitaniae (MICs = 100, 200, and 50 µg/ml, respectively); decreases hepatic and renal injury and increases survival in a mouse model of polyphosphate-induced lethal endotoxemia at 57.6 µg/animal