Learn More
Medchemexpress LLC Luteolin-7-rutinoside | 20633-84-5 | 1 MG
SDP
Catalog No. 5000417995
Encompass_Preferred
Luteolin-7-rutinoside is a flavonoid glycoside exhibiting both anti-arthritic and antifungal activities. It can be used as a combination therapy for the treatment of fungal arthritis due to C. albicans infection. Its suppressive activity is dose-dependent, and it shifts the Th1-Th2 balance away from Th1- towards Th2-type responses, while also inhibiting nitric oxide (NO) production from macrophages.
  • Anti-arthritic activity
  • Antifungal activity
  • Can be used for treatment of fungal arthritis due to C. albicans infection
  • Suppressive activity on T cell proliferation is dose-dependent
  • Shifts Th1-Th2 balance away from Th1- towards Th2-type responses
  • Inhibits nitric oxide (NO) production from macrophages
Catalog No. 50-004-17995 Supplier Medchemexpress LLC Supplier No. HYN66471MG
Luteolin-7-rutinoside is a flavonoid glycoside exhibiting both anti-arthritic and antifungal activities. It can be used as a combination therapy for the treatment of fungal arthritis due to C. albicans infection. Its suppressive activity is dose-dependent, and it shifts the Th1-Th2 balance away from Th1- towards Th2-type responses, while also inhibiting nitric oxide (NO) production from macrophages.
  • Anti-arthritic activity
  • Antifungal activity
  • Can be used for treatment of fungal arthritis due to C. albicans infection
  • Suppressive activity on T cell proliferation is dose-dependent
  • Shifts Th1-Th2 balance away from Th1- towards Th2-type responses
  • Inhibits nitric oxide (NO) production from macrophages