Learn More
Medchemexpress LLC Luteolin-7-rutinoside | 20633-84-5 | 5 MG
SDP
Catalog No. 5000419555
Encompass_Preferred
Luteolin-7-rutinoside possesses both anti-arthritic and antifungal activities, which can lead to a combination therapy for treating fungal arthritis caused by *C. albicans* infection. It is intended for research use only and not for sale to patients.
  • Possesses anti-arthritic activity.
  • Possesses antifungal activity.
  • Reduces proliferation of T cells in a dose-dependent manner.
  • Shifts Th1-Th2 balance away from Th1 towards Th2-type responses.
  • Inhibits nitric oxide (NO) production from macrophages.
Catalog No. 50-004-19555 Supplier Medchemexpress LLC Supplier No. HYN66475MG
Luteolin-7-rutinoside possesses both anti-arthritic and antifungal activities, which can lead to a combination therapy for treating fungal arthritis caused by *C. albicans* infection. It is intended for research use only and not for sale to patients.
  • Possesses anti-arthritic activity.
  • Possesses antifungal activity.
  • Reduces proliferation of T cells in a dose-dependent manner.
  • Shifts Th1-Th2 balance away from Th1 towards Th2-type responses.
  • Inhibits nitric oxide (NO) production from macrophages.